Assay Details
| Gene Symbol: GPR89A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257436.2 | NP_001244365.1 | 217 | 103 | ||
| XM_028829452.1 | XP_028685285.1 | 273 | 103 | |||
| GenBank mRNA | JU334899.1 | - | 218 | 103 | ||
| JU475906.1 | - | 246 | 103 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028829452.1 transcripts.
Set Membership: |
|

GPR89A
Genomic Map