Assay Details
| Gene Symbol: GPR89A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257436.2 | NP_001244365.1 | 845 | 139 | ||
| XM_028829452.1 | XP_028685285.1 | 921 | 139 | |||
| GenBank mRNA | JU334899.1 | - | 846 | 139 | ||
| JU475906.1 | - | 874 | 139 | |||
| JU487935.1 | - | 881 | 139 | |||
More Information
Set Membership: |
|

GPR89A
Genomic Map