Assay Details
| Gene Symbol: HACD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266546.1 | NP_001253475.1 | 354 | 97 | ||
| XM_015142481.2 | XP_014997967.1 | 138 | 97 | |||
| GenBank mRNA | JU322199.1 | - | 349 | 97 | ||
| JU332143.1 | - | 349 | 97 | |||
| JU332144.1 | - | 349 | 97 | |||
| JU473635.1 | - | 354 | 97 | |||
| JU473636.1 | - | 354 | 97 | |||
More Information
Set Membership: |
|

HACD3
Genomic Map