Assay Details
| Gene Symbol: COX16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194897.3 | NP_001181826.2 | 207 | 83 | ||
| GenBank mRNA | JU322906.1 | - | 200 | 83 | ||
| JU475861.1 | - | 191 | 83 | |||
| JV686060.1 | - | 207 | 83 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

COX16
Genomic Map