Assay Details
Gene Symbol: RANGRF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194570.2 | NP_001181499.2 | 585 | 117 | ||
GenBank mRNA | JU322862.1 | - | 558 | 117 | ||
JU470672.1 | - | 580 | 117 | |||
JV045502.1 | - | 559 | 117 | |||
JV668780.1 | - | 556 | 117 | |||
JV703886.1 | - | 706 | 117 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
