Assay Details
| Gene Symbol: PIPOX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015118941.2 | XP_014974427.1 | 1381 | 74 | ||
| XM_015118942.2 | XP_014974428.1 | 1381 | 74 | |||
| GenBank mRNA | JU323352.1 | - | 1024 | 74 | ||
| JU323353.1 | - | 1024 | 74 | |||
| JV704584.1 | - | 1026 | 74 | |||
More Information
Set Membership: |
|

PIPOX
Genomic Map