Assay Details
Gene Symbol: ECSIT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001106441.4 | XP_001106441.2 | 1307 | 95 | ||
XM_015122961.2 | XP_014978447.2 | 1175 | 95 | |||
XM_015122962.2 | XP_014978448.2 | 1200 | 95 | |||
XM_015122964.2 | XP_014978450.2 | 1026 | 95 | |||
XM_015122965.2 | XP_014978451.1 | 533 | 95 | |||
XM_028839111.1 | XP_028694944.1 | 678 | 95 | |||
GenBank mRNA | JU323461.1 | - | 1002 | 95 | ||
JU334848.1 | - | 509 | 95 | |||
JU334849.1 | - | 1151 | 95 | |||
JV047276.1 | - | 488 | 95 | |||
JV047277.1 | - | 1130 | 95 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
