Assay Details
Gene Symbol: RSRC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266753.2 | NP_001253682.1 | 200 | 72 | ||
XM_015131741.2 | XP_014987227.1 | 368 | 72 | |||
XM_015131742.2 | XP_014987228.1 | 337 | 72 | |||
XM_028843222.1 | XP_028699055.1 | 404 | 72 | |||
GenBank mRNA | JU334862.1 | - | 308 | 72 | ||
JU475883.1 | - | 309 | 72 | |||
JV043265.1 | - | 202 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
