Assay Details
Gene Symbol: XAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194803.2 | NP_001181732.1 | 797 | 86 | ||
XM_001103678.4 | XP_001103678.1 | 1718 | 86 | |||
XM_028835641.1 | XP_028691474.1 | 1635 | 86 | |||
XM_028835643.1 | XP_028691476.1 | 1578 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
