Assay Details
Gene Symbol: CNNM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028832180.1 | XP_028688013.1 | 4039 | 93 | ||
XM_028832182.1 | XP_028688015.1 | 4039 | 93 | |||
XM_028832183.1 | XP_028688016.1 | 3967 | 93 | |||
XM_028832184.1 | XP_028688017.1 | 3817 | 93 | |||
XM_028832185.1 | XP_028688018.1 | 4039 | 93 | |||
GenBank mRNA | JU326536.1 | - | 1085 | 93 | ||
JU484381.1 | - | 1097 | 93 | |||
JV711899.1 | - | 1033 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
