Assay Details
| Gene Symbol: GIPC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001104248.4 | XP_001104248.1 | 984 | 127 | ||
| XM_015143166.2 | XP_014998652.2 | 996 | 127 | |||
| XM_015143174.2 | XP_014998660.1 | 1150 | 127 | |||
More Information
Set Membership: |
|

GIPC2
Genomic Map