Assay Details
Gene Symbol: RETSAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257563.1 | NP_001244492.1 | 308 | 83 | ||
GenBank mRNA | JU323377.1 | - | 308 | 83 | ||
JU473683.1 | - | 355 | 83 | |||
JV047464.1 | - | 312 | 83 | |||
JV529303.1 | - | 205 | 83 | |||
JV638595.1 | - | 364 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() |
