Assay Details
| Gene Symbol: WBP1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194587.3 | NP_001181516.1 | 388 | 92 | ||
| XM_001113270.4 | XP_001113270.1 | 1517 | 92 | |||
| XM_015148010.2 | XP_015003496.1 | 1389 | 92 | |||
| XM_028853499.1 | XP_028709332.1 | 277 | 92 | |||
| GenBank mRNA | JV585448.1 | - | 439 | 92 | ||
| JV635688.1 | - | 358 | 92 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028853499.1 transcripts.
Set Membership: |
|

WBP1L
Genomic Map