Assay Details
Gene Symbol: BPNT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257950.1 | NP_001244879.1 | 592 | 152 | ||
GenBank mRNA | JU321489.1 | - | 592 | 152 | ||
JU332180.1 | - | 592 | 152 | |||
JU473746.1 | - | 630 | 152 | |||
JV511129.1 | - | 376 | 152 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
