Assay Details
| Gene Symbol: C1H1orf109 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028835816.1 | XP_028691649.1 | 1533 | 116 | ||
| XM_028835817.1 | XP_028691650.1 | 1134 | 116 | |||
| XM_028835822.1 | XP_028691655.1 | 834 | 116 | |||
| XM_028835828.1 | XP_028691661.1 | 797 | 116 | |||
| XM_028835835.1 | XP_028691668.1 | 832 | 116 | |||
| GenBank mRNA | JU326099.1 | - | 442 | 116 | ||
More Information
Set Membership: |
|

C1H1orf109
Genomic Map