Assay Details
Gene Symbol: THG1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194604.2 | NP_001181533.2 | 856 | 86 | ||
XM_015141291.2 | XP_014996777.2 | 789 | 86 | |||
XM_015141292.2 | XP_014996778.2 | 652 | 86 | |||
XM_028849280.1 | XP_028705113.1 | 683 | 86 | |||
XM_028849281.1 | XP_028705114.1 | 338 | 86 | |||
GenBank mRNA | JU476165.1 | - | 760 | 86 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
