Assay Details
Gene Symbol: MAGOHB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265616.1 | NP_001252545.1 | 314 | 138 | ||
GenBank mRNA | JU328199.1 | - | 244 | 138 | ||
JV064694.1 | - | 316 | 138 | |||
JV636250.1 | - | 314 | 138 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
