Assay Details
Gene Symbol: LOC717842 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001116222.4 | XP_001116222.2 | 1942 | 119 | ||
XM_028828102.1 | XP_028683935.1 | 1310 | 119 |
Gene Symbol: LOC717923 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JV585475.1 | - | 3411 | 119 | ||
JV717626.1 | - | 3410 | 119 |

More Information
Set Membership: |
![]() ![]() ![]() |
