Assay Details
Gene Symbol: LOC717923 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JV585475.1 | - | 4495 | 111 | ||
JV717626.1 | - | 4494 | 111 |
Gene Symbol: LOC717842 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001116222.4 | XP_001116222.2 | 858 | 111 | ||
XM_028828085.1 | XP_028683918.1 | 858 | 111 | |||
XM_028828090.1 | XP_028683923.1 | 858 | 111 | |||
XM_028828096.1 | XP_028683929.1 | 858 | 111 | |||
XM_028828100.1 | XP_028683933.1 | 537 | 111 | |||
XM_028828102.1 | XP_028683935.1 | 226 | 111 | |||
XM_028828106.1 | XP_028683939.1 | 858 | 111 | |||
GenBank mRNA | JV064751.1 | - | 402 | 111 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028828100.1, XM_028828102.1 transcripts.
Set Membership: |
![]() ![]() |
