Assay Details
| Gene Symbol: CKAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001084035.4 | XP_001084035.2 | 1612 | 109 | ||
| XM_015121139.2 | XP_014976625.2 | 1674 | 109 | |||
| XM_015121140.2 | XP_014976626.2 | 1623 | 109 | |||
| XM_015121141.2 | XP_014976627.2 | 1590 | 109 | |||
| GenBank mRNA | JU484400.1 | - | 1566 | 109 | ||
More Information
Set Membership: |
|

CKAP2
Genomic Map