Assay Details
| Gene Symbol: LOC703026 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015133369.2 | XP_014988855.1 | 549 | 188 | ||
| XM_028845708.1 | XP_028701541.1 | 549 | 188 | |||
| GenBank mRNA | JU335288.1 | - | 518 | 188 | ||
| JU476247.1 | - | 381 | 188 | |||
| JV047535.1 | - | 414 | 188 | |||
| JV636252.1 | - | 506 | 188 | |||
More Information
Set Membership: |
|

LOC703026
Genomic Map