Assay Details
Gene Symbol: LOC703026 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015133369.2 | XP_014988855.1 | 1155 | 149 | ||
XM_028845708.1 | XP_028701541.1 | 1155 | 149 | |||
GenBank mRNA | JU335288.1 | - | 1124 | 149 | ||
JU476247.1 | - | 987 | 149 | |||
JV047535.1 | - | 1020 | 149 | |||
JV636252.1 | - | 1112 | 149 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
