Assay Details
Gene Symbol: LRRC36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001089982.4 | XP_001089982.1 | 534 | 93 | ||
XM_015126388.2 | XP_014981874.1 | 512 | 93 | |||
XM_015126389.2 | XP_014981875.1 | 512 | 93 | |||
XM_015126390.2 | XP_014981876.1 | 511 | 93 | |||
XM_015126392.2 | XP_014981878.1 | 518 | 93 | |||
XM_028840785.1 | XP_028696618.1 | 512 | 93 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015126392.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
