Assay Details
| Gene Symbol: RNF121 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194615.3 | NP_001181544.1 | 874 | 77 | ||
| GenBank mRNA | JV600846.1 | - | 862 | 77 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RNF121| Gene Symbol: RNF121 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194615.3 | NP_001181544.1 | 874 | 77 | ||
| GenBank mRNA | JV600846.1 | - | 862 | 77 | ||
Set Membership: |
|