Assay Details
| Gene Symbol: RIOK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260597.1 | NP_001247526.1 | 132 | 66 | ||
| GenBank mRNA | JU335430.1 | - | 76 | 66 | ||
| JU476374.1 | - | 132 | 66 | |||
More Information
Set Membership: |
|

RIOK2
Genomic Map