Assay Details
Gene Symbol: CDCA7L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257527.1 | NP_001244456.1 | 430 | 86 | ||
XM_015133877.1 | XP_014989363.1 | 582 | 86 | |||
XM_015133878.2 | XP_014989364.1 | 420 | 86 | |||
XM_015133879.1 | XP_014989365.1 | 418 | 86 | |||
XM_028844996.1 | XP_028700829.1 | 582 | 86 | |||
GenBank mRNA | JU335344.1 | - | 430 | 86 | ||
JU476296.1 | - | 414 | 86 | |||
JU476297.1 | - | 414 | 86 | |||
JU476298.1 | - | 414 | 86 | |||
JV047577.1 | - | 389 | 86 | |||
JV047578.1 | - | 389 | 86 | |||
JV047579.1 | - | 389 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() |
