Assay Details
Gene Symbol: CCSER2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193762.2 | NP_001180691.2 | 2067 | 124 | ||
XM_015147096.2 | XP_015002582.2 | 2154 | 124 | |||
XM_015147097.2 | XP_015002583.2 | 2005 | 124 | |||
XM_015147098.2 | XP_015002584.2 | 2154 | 124 | |||
XM_015147099.2 | XP_015002585.2 | 2154 | 124 | |||
XM_015147100.2 | XP_015002586.2 | 2154 | 124 | |||
XM_015147101.2 | XP_015002587.1 | 350 | 124 | |||
XM_015147104.2 | XP_015002590.1 | 311 | 124 | |||
XM_015147106.2 | XP_015002592.1 | 371 | 124 | |||
XM_015147107.2 | XP_015002593.1 | 371 | 124 |

More Information
Set Membership: |
![]() ![]() ![]() |
