Assay Details
Gene Symbol: SEPTIN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015150480.2 | XP_015005966.1 | 638 | 143 | ||
XM_015150481.2 | XP_015005967.1 | 663 | 143 | |||
XM_015150482.2 | XP_015005968.1 | 726 | 143 | |||
XM_028827647.1 | XP_028683480.1 | 1926 | 143 | |||
XM_028827648.1 | XP_028683481.1 | 1926 | 143 | |||
XM_028827649.1 | XP_028683482.1 | 683 | 143 | |||
XM_028827650.1 | XP_028683483.1 | 844 | 143 | |||
GenBank mRNA | JU319811.1 | - | 546 | 143 | ||
JV587371.1 | - | 557 | 143 | |||
JV587372.1 | - | 557 | 143 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
