Assay Details
| Gene Symbol: POLE4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261810.1 | NP_001248739.1 | 320 | 112 | ||
| GenBank mRNA | JU321789.1 | - | 319 | 112 | ||
| JU473370.1 | - | 320 | 112 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

POLE4
Genomic Map