Assay Details
| Gene Symbol: YAE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194130.2 | NP_001181059.2 | 181 | 151 | ||
| GenBank mRNA | JU323116.1 | - | 146 | 151 | ||
| JV043799.1 | - | 141 | 151 | |||
| JV567130.1 | - | 134 | 151 | |||
More Information
Set Membership: |
|

YAE1
Genomic Map