Assay Details
| Gene Symbol: CASS4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015148735.2 | XP_015004221.2 | 2904 | 120 | ||
| XM_028828521.1 | XP_028684354.1 | 2254 | 120 | |||
More Information
Set Membership: |
|

CASS4
Genomic Map