Assay Details
| Gene Symbol: SALL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001093669.4 | XP_001093669.1 | 592 | 125 | ||
| XM_028828514.1 | XP_028684347.1 | 588 | 125 | |||
| GenBank mRNA | JV656626.1 | - | 80 | 125 | ||
More Information
Set Membership: |
|

SALL4
Genomic Map