Assay Details
Gene Symbol: LOC715257 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015110742.2 | XP_014966228.2 | 1112 | 158 | ||
XM_015110743.2 | XP_014966229.2 | 1152 | 158 | |||
XM_015110744.2 | XP_014966230.2 | 975 | 158 | |||
XM_015110745.2 | XP_014966231.2 | 854 | 158 | |||
XM_028831002.1 | XP_028686835.1 | 1395 | 158 | |||
XM_028831003.1 | XP_028686836.1 | 1000 | 158 | |||
GenBank mRNA | JU326862.1 | - | 760 | 158 | ||
JU476529.1 | - | 885 | 158 | |||
JV635566.1 | - | 1419 | 158 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
