Assay Details
| Gene Symbol: ZNF248 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001095695.4 | XP_001095695.1 | 899 | 109 | ||
| XM_015146858.2 | XP_015002344.1 | 853 | 109 | |||
| XM_028826214.1 | XP_028682047.1 | 242 | 109 | |||
| XM_028826217.1 | XP_028682050.1 | 242 | 109 | |||
| GenBank mRNA | JU335598.1 | - | 509 | 109 | ||
| JV719321.1 | - | 514 | 109 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001095695.4, XM_015146858.2, XM_028826214.1, XM_028826217.1 transcripts.
Set Membership: |
|

ZNF248
Genomic Map