Assay Details
| Gene Symbol: ENPP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015136489.2 | XP_014991975.1 | 1252 | 111 | ||
| XM_015136490.2 | XP_014991976.1 | 1306 | 111 | |||
| XM_015136491.2 | XP_014991977.1 | 1126 | 111 | |||
| XM_015136492.2 | XP_014991978.1 | 1107 | 111 | |||
| GenBank mRNA | JU323211.1 | - | 1206 | 111 | ||
| JU472790.1 | - | 1049 | 111 | |||
More Information
Set Membership: |
|

ENPP5
Genomic Map