Assay Details
Gene Symbol: FCGR2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257300.1 | NP_001244229.1 | 82 | 112 | ||
XM_015113136.2 | XP_014968622.2 | 143 | 115 | |||
XM_028846295.1 | XP_028702128.1 | 141 | 115 | |||
XM_028846297.1 | XP_028702130.1 | 157 | 112 | |||
GenBank mRNA | JQ037997.1 | - | 82 | 112 | ||
JQ037998.2 | - | 82 | 112 | |||
JQ037999.1 | - | 82 | 112 | |||
JQ038000.2 | - | 82 | 112 | |||
MK318564.1 | - | 82 | 115 | |||
MK318565.1 | - | 82 | 112 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
