Assay Details
| Gene Symbol: POPDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194472.3 | NP_001181401.2 | 771 | 131 | ||
| XM_015129907.2 | XP_014985393.2 | 1059 | 131 | |||
| XM_015129908.2 | XP_014985394.2 | 1060 | 131 | |||
| GenBank mRNA | JV658565.1 | - | 137 | 131 | ||
More Information
Set Membership: |
|

POPDC2
Genomic Map