Assay Details
| Gene Symbol: MRPL46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001088970.4 | XP_001088970.2 | 652 | 162 | ||
| GenBank mRNA | JU323426.1 | - | 162 | |||
| JU473064.1 | - | 162 | ||||
| JV045790.1 | - | 162 | ||||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

MRPL46
Genomic Map