Assay Details
| Gene Symbol: MRPS11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261578.2 | NP_001248507.1 | 297 | 150 | ||
| XM_015142938.2 | XP_014998424.1 | 489 | 150 | |||
| XM_015142939.2 | XP_014998425.1 | 1427 | 150 | |||
| GenBank mRNA | JU335969.1 | - | 293 | 150 | ||
| JU476852.1 | - | 297 | 150 | |||
| JV659122.1 | - | 297 | 150 | |||
More Information
Set Membership: |
|

MRPS11
Genomic Map