Assay Details
| Gene Symbol: GRAMD2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265990.1 | NP_001252919.1 | 577 | 129 | ||
| XM_015140671.2 | XP_014996157.1 | 545 | 129 | |||
| XM_015140672.2 | XP_014996158.1 | 888 | 129 | |||
| XM_028849172.1 | XP_028705005.1 | 545 | 129 | |||
| XM_028849173.1 | XP_028705006.1 | 888 | 129 | |||
| XM_028849174.1 | XP_028705007.1 | 845 | 129 | |||
| XM_028849175.1 | XP_028705008.1 | 578 | 129 | |||
| XM_028849177.1 | XP_028705010.1 | 1584 | 129 | |||
| GenBank mRNA | JU321037.1 | - | 577 | 129 | ||
| JU322547.1 | - | 620 | 129 | |||
More Information
Set Membership: |
|

GRAMD2B
Genomic Map