Assay Details
Gene Symbol: LOC106993830 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028842748.1 | XP_028698581.1 | 1084 | 132 | ||
XM_028842749.1 | XP_028698582.1 | 1059 | 132 | |||
XM_028842750.1 | XP_028698583.1 | 1059 | 132 | |||
XM_028842751.1 | XP_028698584.1 | 1059 | 132 | |||
XM_028842752.1 | XP_028698585.1 | 1059 | 132 |
Gene Symbol: SECISBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001090428.4 | XP_001090428.2 | 2259 | 132 | ||
XM_001090657.3 | XP_001090657.2 | 2375 | 132 | |||
XM_015118054.2 | XP_014973540.2 | 2260 | 132 | |||
XM_015118055.2 | XP_014973541.2 | 2257 | 132 | |||
XM_015118058.2 | XP_014973544.2 | 2792 | 132 | |||
XM_028835315.1 | XP_028691148.1 | 2375 | 132 | |||
XM_028835316.1 | XP_028691149.1 | 2913 | 132 | |||
XM_028835317.1 | XP_028691150.1 | 2652 | 132 | |||
GenBank mRNA | JU336342.1 | - | 2348 | 132 | ||
JV723171.1 | - | 2350 | 132 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() |
