Assay Details
| Gene Symbol: NOC4L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258179.1 | NP_001245108.1 | 1367 | 77 | ||
| XM_015109722.2 | XP_014965208.2 | 1356 | 77 | |||
| XM_015109723.2 | XP_014965209.2 | 3359 | 77 | |||
| XM_015109724.2 | XP_014965210.2 | 3141 | 77 | |||
| XM_028828935.1 | XP_028684768.1 | 2867 | 77 | |||
| GenBank mRNA | JU323182.1 | - | 1350 | 77 | ||
| JU473445.1 | - | 1367 | 77 | |||
| JV045522.1 | - | 1363 | 77 | |||
| JV565185.1 | - | 1367 | 77 | |||
| JV661177.1 | - | 1293 | 77 | |||
More Information
Set Membership: |
|

NOC4L
Genomic Map