Assay Details
| Gene Symbol: TRAPPC6A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194478.3 | NP_001181407.2 | 321 | 141 | ||
| XM_002801293.3 | XP_002801339.2 | 332 | 141 | |||
| XM_015124121.2 | XP_014979607.2 | 650 | 141 | |||
| GenBank mRNA | JU493294.1 | - | 185 | 141 | ||
More Information
Set Membership: |
|

TRAPPC6A
Genomic Map