Assay Details
| Gene Symbol: RETREG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193956.2 | NP_001180885.2 | 416 | 122 | ||
| XM_028830502.1 | XP_028686335.1 | 411 | 122 | |||
| GenBank mRNA | JU326611.1 | - | 268 | 122 | ||
| JV527814.1 | - | 291 | 122 | |||
| JV557589.1 | - | 291 | 122 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028830502.1 transcripts.
Set Membership: |
|

RETREG2
Genomic Map