Assay Details
Gene Symbol: RETREG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193956.2 | NP_001180885.2 | 1008 | 141 | ||
XM_028830502.1 | XP_028686335.1 | 915 | 141 | |||
GenBank mRNA | JU326611.1 | - | 860 | 141 | ||
JV527814.1 | - | 883 | 141 | |||
JV557589.1 | - | 883 | 141 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() |
