Assay Details
Gene Symbol: QSER1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257718.1 | NP_001244647.1 | 80 | 84 | ||
XM_015114500.2 | XP_014969986.1 | 1231 | 84 | |||
XM_028832510.1 | XP_028688343.1 | 332 | 84 | |||
GenBank mRNA | JU336429.1 | - | 80 | 84 | ||
JU477247.1 | - | 125 | 84 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001257718.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
