Assay Details
Gene Symbol: TLCD3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028835838.1 | XP_028691671.1 | 232 | 50 | ||
XM_028835839.1 | XP_028691672.1 | 407 | 50 | |||
GenBank mRNA | JV594057.1 | - | 125 | 50 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
