Assay Details
Gene Symbol: STN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266590.1 | NP_001253519.1 | 998 | 106 | ||
XM_028853528.1 | XP_028709361.1 | 1129 | 106 | |||
XM_028853529.1 | XP_028709362.1 | 1357 | 106 | |||
XM_028853530.1 | XP_028709363.1 | 1067 | 106 | |||
XM_028853531.1 | XP_028709364.1 | 1430 | 106 | |||
XM_028853532.1 | XP_028709365.1 | 1129 | 106 | |||
XM_028853533.1 | XP_028709366.1 | 1129 | 106 | |||
GenBank mRNA | JU322960.1 | - | 1027 | 106 | ||
JU477278.1 | - | 918 | 106 | |||
JV636384.1 | - | 998 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() |
