Assay Details
| Gene Symbol: SIKE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261494.1 | NP_001248423.1 | 473 | 104 | ||
| XM_015147217.2 | XP_015002703.1 | 512 | 104 | |||
| GenBank mRNA | JU336479.1 | - | 470 | 104 | ||
| JU477288.1 | - | 473 | 104 | |||
More Information
Set Membership: |
|

SIKE1
Genomic Map