Assay Details
| Gene Symbol: ASRGL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194716.3 | NP_001181645.1 | 300 | 73 | ||
| GenBank mRNA | JU323185.1 | - | 321 | 73 | ||
| JU323186.1 | - | 321 | 73 | |||
| JV045810.1 | - | 317 | 73 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

ASRGL1
Genomic Map